This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03230689_m1
See other UBE2L3 GE Assays ›
Gene Symbol:
UBE2L3
Gene Name:
ubiquitin conjugating enzyme E2 L3
Gene Aliases:
-
Chromosome Location:
Chr.17: 72128567 - 72153607 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03230689_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBE2L3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001078094.1 NP_001071562.1 2 - 3 161 87
GenBank mRNA BC105164.1 - 2 - 3 161 87
BT025380.1 - 2 - 3 121 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
regulation of transcription, DNA-templated
cell proliferation
protein ubiquitination
cell cycle phase transition
protein K11-linked ubiquitination
cellular response to steroid hormone stimulus
cellular response to glucocorticoid stimulus
positive regulation of nucleic acid-templated transcription
transcription coactivator activity
ubiquitin-protein transferase activity
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0