Assay Details
| Gene Symbol: GTSF1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079601.2 | NP_001073069.1 | 1 | 851 | 75 | |
| GenBank mRNA | BC102599.1 | - | 1 | 852 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GTSF1L| Gene Symbol: GTSF1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079601.2 | NP_001073069.1 | 1 | 851 | 75 | |
| GenBank mRNA | BC102599.1 | - | 1 | 852 | 75 | |
Set Membership: |
|