Assay Details
| Gene Symbol: BIRC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001855.3 | NP_001001855.2 | 2 - 3 | 291 | 71 | |
| GenBank mRNA | BC109566.1 | - | 2 - 3 | 292 | 71 | |
More Information
Set Membership: |
|

BIRC5
Genomic Map