This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03232303_m1
See other TRAPPC6B GE Assays ›
Gene Symbol:
TRAPPC6B
Gene Name:
trafficking protein particle complex 6B
Gene Aliases:
-
Chromosome Location:
Chr.21: 49145029 - 49156067 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03232303_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRAPPC6B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001038134.1 NP_001033223.1 1 - 2 231 93
GenBank mRNA BC109723.1 - 1 - 2 231 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons

Gene Ontology Categories:

Function(s)

nervous system development
regulation of GTPase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline