This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03232818_m1
See other NTMT1 GE Assays ›
Gene Symbol:
NTMT1
Gene Name:
N-terminal Xaa-Pro-Lys N-methyltransferase 1
Gene Aliases:
C11H9orf32, C9orf32, METTL11A, NTM1A
Chromosome Location:
Chr.11: 100016861 - 100029018 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03232818_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NTMT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001038217.2 NP_001033306.1 2 - 3 360 64
GenBank mRNA BC111344.1 - 2 - 3 361 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

spindle organization
chromosome segregation
histone methylation
N-terminal peptidyl-glycine methylation
N-terminal peptidyl-proline dimethylation
N-terminal peptidyl-serine dimethylation
N-terminal peptidyl-serine trimethylation
protein methyltransferase activity
histone methyltransferase activity
N-terminal protein N-methyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline