This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03235183_m1
See other KMT5A GE Assays ›
Gene Symbol:
KMT5A
Gene Name:
lysine methyltransferase 5A
Gene Aliases:
SETD8
Chromosome Location:
Chr.17: 52210477 - 52228187 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03235183_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KMT5A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001046330.1 NP_001039795.1 3 - 4 346 65
GenBank mRNA BC110189.1 - 3 - 4 346 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
cell cycle
peptidyl-lysine monomethylation
histone lysine methylation
regulation of DNA damage response, signal transduction by p53 class mediator
cell division
p53 binding
transcription corepressor activity
protein-lysine N-methyltransferase activity
histone-lysine N-methyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6dxlp:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0