This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03236071_m1
See other PDGFA GE Assays ›
Gene Symbol:
PDGFA
Gene Name:
platelet derived growth factor subunit A
Gene Aliases:
-
Chromosome Location:
Chr.25: 42012070 - 42031485 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03236071_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDGFA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001075231.1 NP_001068699.1 3 - 4 349 70
GenBank mRNA BC105569.1 - 3 - 4 349 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
positive regulation of cell proliferation
response to wounding
negative regulation of phosphatidylinositol biosynthetic process
negative regulation of platelet activation
positive regulation of phosphatidylinositol 3-kinase signaling
regulation of smooth muscle cell migration
positive regulation of cell migration
positive regulation of protein autophosphorylation
positive regulation of metanephric mesenchymal cell migration by platelet-derived growth factor receptor-beta signaling pathway
positive regulation of MAP kinase activity
positive regulation of MAPK cascade
platelet-derived growth factor receptor signaling pathway
positive regulation of fibroblast proliferation
negative chemotaxis
positive regulation of cell division
positive regulation of protein kinase B signaling
positive regulation of ERK1 and ERK2 cascade
regulation of glomerular mesangial cell proliferation
regulation of DNA biosynthetic process
platelet-derived growth factor receptor binding
collagen binding
growth factor activity
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline