This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03237073_m1
See other TRAPPC5 GE Assays ›
Gene Symbol:
TRAPPC5
Gene Name:
trafficking protein particle complex 5
Gene Aliases:
-
Chromosome Location:
Chr.7: 16526003 - 16528272 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03237073_m1
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRAPPC5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001077849.2 NP_001071317.1 1 - 2 35 75
GenBank mRNA BC111232.1 - 1 - 2 36 75

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC111232.1, NM_001077849.2 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Inventoried Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0