This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03237629_m1
See other PPP2R2B GE Assays ›
Gene Symbol:
PPP2R2B
Gene Name:
protein phosphatase 2 regulatory subunit Bbeta
Gene Aliases:
-
Chromosome Location:
Chr.7: 57796194 - 58305719 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03237629_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP2R2B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001272085.1 NP_001259014.1 8 - 9 1092 62
NR_073586.1 7 - 8 1413 62
GenBank mRNA BC118167.1 - 7 - 8 1413 62
BT020863.1 - 7 - 8 1093 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s)

regulation of phosphoprotein phosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nbrrd:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0