This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03239818_m1
See other CALHM2 GE Assays ›
Gene Symbol:
CALHM2
Gene Name:
calcium homeostasis modulator family member 2
Gene Aliases:
-
Chromosome Location:
Chr.26: 24030502 - 24035854 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03239818_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CALHM2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001046420.1 NP_001039885.1 1 - 2 249 79
GenBank mRNA BC113290.1 - 1 - 2 249 79

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC113290.1, NM_001046420.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons

Gene Ontology Categories:

Function(s)

positive regulation of apoptotic process
cation transmembrane transport

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline