This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03240343_m1
See other CISH GE Assays ›
Gene Symbol:
CISH
Gene Name:
cytokine inducible SH2 containing protein
Gene Aliases:
-
Chromosome Location:
Chr.22: 49750073 - 49755491 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03240343_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CISH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001046586.1 NP_001040051.1 1 - 2 120 54
GenBank mRNA BC113307.1 - 1 - 2 120 54

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC113307.1, NM_001046586.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

kinase modulator

Gene Ontology Categories:

Function(s)

protein kinase C-activating G-protein coupled receptor signaling pathway
negative regulation of signal transduction
protein ubiquitination
intracellular signal transduction
regulation of growth
regulation of phosphatidylinositol 3-kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline