This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03240493_m1
See other LPL GE Assays ›
Gene Symbol:
LPL
Gene Name:
lipoprotein lipase
Gene Aliases:
-
Chromosome Location:
Chr.8: 66989665 - 67016694 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03240493_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LPL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001075120.1 NP_001068588.1 3 - 4 613 78
GenBank mRNA BC118091.1 - 3 - 4 613 78
M16966.1 - 2 - 3 362 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

fatty acid biosynthetic process
triglyceride metabolic process
phospholipid metabolic process
response to bacterium
response to glucose
positive regulation of macrophage derived foam cell differentiation
positive regulation of cholesterol storage
positive regulation of sequestering of triglyceride
triglyceride catabolic process
cellular response to nutrient
chylomicron remodeling
very-low-density lipoprotein particle remodeling
cholesterol homeostasis
positive regulation of fat cell differentiation
positive regulation of inflammatory response
low-density lipoprotein particle mediated signaling
triglyceride homeostasis
cellular response to fatty acid
positive regulation of chemokine secretion
positive regulation of tumor necrosis factor secretion
lipoprotein lipase activity
phospholipase activity
triglyceride lipase activity
receptor binding
calcium ion binding
protein binding
heparin binding
phosphatidylcholine 1-acylhydrolase activity
apolipoprotein binding
protein homodimerization activity
heparan sulfate proteoglycan binding
phosphatidylserine 1-acylhydrolase activity
1-acyl-2-lysophosphatidylserine acylhydrolase activity
lipoprotein particle binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0