This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03241744_m1
See other NAA60 GE Assays ›
Gene Symbol:
NAA60
Gene Name:
N-alpha-acetyltransferase 60, NatF catalytic subunit
Gene Aliases:
HAT4, NAT15, NatF
Chromosome Location:
Chr.25: 2871247 - 2888334 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03241744_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NAA60
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001075649.1 NP_001069117.1 6 - 7 1085 72
GenBank mRNA BC118298.1 - 6 - 7 1085 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

acetyltransferase

Gene Ontology Categories:

Function(s) Process(es)

nucleosome assembly
N-terminal protein amino acid acetylation
chromosome segregation
cell proliferation
N-terminal peptidyl-methionine acetylation
histone H4 acetylation
peptide alpha-N-acetyltransferase activity
H4 histone acetyltransferase activity
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0