This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03241891_m1
See other UBE3D GE Assays ›
Gene Symbol:
UBE3D
Gene Name:
ubiquitin protein ligase E3D
Gene Aliases:
UBE2CBP
Chromosome Location:
Chr.9: 22779438 - 22958591 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03241891_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBE3D
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001075711.1 NP_001069179.1 7 - 8 885 131
GenBank mRNA BC116118.1 - 7 - 8 885 131

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase protein modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0