This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03242834_m1
See other UGT3A2 GE Assays ›
Gene Symbol:
UGT3A2
Gene Name:
UDP glycosyltransferase family 3 member A2
Gene Aliases:
UGT3A1
Chromosome Location:
Chr.20: 38154289 - 38179041 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03242834_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UGT3A2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001076087.1 NP_001069555.1 2 - 3 295 94
GenBank mRNA BC115988.1 - 2 - 3 295 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons

Gene Ontology Categories:

Process(es)

glucuronosyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0