This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03244351_m1
See other GJA1 GE Assays ›
Gene Symbol:
GJA1
Gene Name:
gap junction protein alpha 1
Gene Aliases:
-
Chromosome Location:
Chr.9: 29748655 - 29761667 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03244351_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GJA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_174068.2 NP_776493.1 1 - 2 190 67
GenBank mRNA BC105464.1 - 1 - 2 190 67

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC105464.1, NM_174068.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

gap junction

Gene Ontology Categories:

Function(s) Process(es)

establishment of mitotic spindle orientation
in utero embryonic development
neuron migration
heart looping
epithelial cell maturation
cell communication
signal transduction
cell-cell signaling
spermatogenesis
heart development
adult heart development
regulation of heart contraction
negative regulation of cell proliferation
response to pH
cell communication by electrical coupling
ATP transport
negative regulation of cell growth
negative regulation of gonadotropin secretion
ion transmembrane transport
cellular protein localization
embryonic heart tube development
regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
skeletal muscle tissue regeneration
positive regulation of protein catabolic process
positive regulation of striated muscle tissue development
bone remodeling
blood vessel morphogenesis
neuron projection morphogenesis
protein oligomerization
bone development
microtubule-based transport
negative regulation of trophoblast cell migration
positive regulation of morphogenesis of an epithelium
positive regulation of mesodermal cell differentiation
positive regulation of stem cell proliferation
gap junction channel activity
beta-catenin binding
ion transmembrane transporter activity
tubulin binding
SH3 domain binding
PDZ domain binding
gap junction hemi-channel activity
gap junction channel activity involved in cell communication by electrical coupling

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline