Assay Details
| Gene Symbol: PPID | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174153.3 | NP_776578.1 | 1 - 2 | 160 | 114 | |
| GenBank mRNA | AB099054.1 | - | 1 - 2 | 153 | 115 | |
| BC113318.1 | - | 1 - 2 | 160 | 114 | ||
| D14074.1 | - | 1 - 2 | 207 | 114 | ||
More Information
Set Membership: |
|

PPID
Genomic Map