This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03244361_m1
See other PPID GE Assays ›
Gene Symbol:
PPID
Gene Name:
peptidylprolyl isomerase D
Gene Aliases:
CYP-40
Chromosome Location:
Chr.17: 40279452 - 40292371 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03244361_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPID
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_174153.3 NP_776578.1 2 - 3 289 84
GenBank mRNA BC113318.1 - 2 - 3 289 84
D14074.1 - 2 - 3 336 84
L11668.1 - 1 - 2 107 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
protein peptidyl-prolyl isomerization
protein folding
apoptotic process
protein transport
lipid particle organization
positive regulation of apoptotic process
positive regulation of viral genome replication
positive regulation of protein secretion
chaperone-mediated protein folding
protein-containing complex assembly
cellular response to UV-A
peptidyl-prolyl cis-trans isomerase activity
protein binding
transcription factor binding
cyclosporin A binding
estrogen receptor binding
Hsp70 protein binding
Hsp90 protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline