This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03244363_g1
See other PPID GE Assays ›
Gene Symbol:
PPID
Gene Name:
peptidylprolyl isomerase D
Gene Aliases:
CYP-40
Chromosome Location:
Chr.17: 40279452 - 40292371 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03244363_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPID
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_174153.3 NP_776578.1 4 - 5 596 84
GenBank mRNA BC113318.1 - 4 - 5 596 84
D14074.1 - 4 - 5 643 84
L11668.1 - 3 - 4 414 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
protein peptidyl-prolyl isomerization
protein folding
apoptotic process
protein transport
lipid particle organization
positive regulation of apoptotic process
positive regulation of viral genome replication
positive regulation of protein secretion
chaperone-mediated protein folding
protein-containing complex assembly
cellular response to UV-A
peptidyl-prolyl cis-trans isomerase activity
protein binding
transcription factor binding
cyclosporin A binding
estrogen receptor binding
Hsp70 protein binding
Hsp90 protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0