This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03244581_m1
See other BCAT2 GE Assays ›
Gene Symbol:
BCAT2
Gene Name:
branched chain amino acid transaminase 2
Gene Aliases:
-
Chromosome Location:
Chr.18: 55411008 - 55425020 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03244581_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BCAT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013593.2 NP_001013611.2 6 - 7 705 100
GenBank mRNA BC114055.1 - 6 - 7 721 100
BT020729.1 - 6 - 7 826 100
BT020823.1 - 6 - 7 734 100
BT021163.1 - 6 - 7 705 100
BT026175.1 - 6 - 7 705 100
BT026222.1 - 5 - 6 688 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

transaminase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Process(es)

L-leucine transaminase activity
L-valine transaminase activity
L-isoleucine transaminase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline