This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03247571_m1
See other MOV10 GE Assays ›
Gene Symbol:
MOV10
Gene Name:
Mov10 RISC complex RNA helicase
Gene Aliases:
-
Chromosome Location:
Chr.3: 30624834 - 30652044 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03247571_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MOV10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001075839.1 NP_001069307.1 6 - 7 1057 61
GenBank mRNA BC148151.1 - 6 - 7 1058 61
BT026242.1 - 6 - 7 1057 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transposition, RNA-mediated
gene silencing by miRNA
mRNA cleavage involved in gene silencing by miRNA
defense response to virus
positive regulation of mRNA catabolic process
3'-UTR-mediated mRNA destabilization
regulation of neuron projection arborization
RNA binding
ATP binding
5'-3' RNA helicase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0