Assay Details
| Gene Symbol: CAMKV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075876.1 | NP_001069344.1 | 8 - 9 | 831 | 70 | |
| GenBank mRNA | BC123694.1 | - | 8 - 9 | 831 | 70 | |
More Information
Set Membership: |
|

CAMKV
Genomic Map