This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03248753_m1
See other MLLT11 GE Assays ›
Gene Symbol:
MLLT11
Gene Name:
MLLT11 transcription factor 7 cofactor
Gene Aliases:
-
Chromosome Location:
Chr.3: 19721036 - 19730753 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03248753_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MLLT11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001076169.1 NP_001069637.1 1 - 2 60 86
GenBank mRNA BC120019.1 - 1 - 2 60 86

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC120019.1, NM_001076169.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons

Gene Ontology Categories:

Function(s)

positive regulation of apoptotic process
positive regulation of transcription, DNA-templated
positive regulation of mitochondrial depolarization
positive regulation of release of cytochrome c from mitochondria
extrinsic apoptotic signaling pathway
intrinsic apoptotic signaling pathway

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0