Assay Details
| Gene Symbol: PJA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080366.1 | NP_001073835.1 | 1 - 2 | 83 | 84 | |
| GenBank mRNA | BT026528.1 | - | 1 - 2 | 83 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001080366.1 transcripts.
Set Membership: |
|

PJA1
Genomic Map