This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03251878_m1
See other NFE2L2 GE Assays ›
Gene Symbol:
NFE2L2
Gene Name:
nuclear factor, erythroid 2 like 2
Gene Aliases:
NRF2
Chromosome Location:
Chr.2: 19468309 - 19649655 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03251878_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 330.60
330.6
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NFE2L2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001011678.2 NP_001011678.2 2 - 3 401 74
GenBank mRNA AB162435.1 - 2 - 3 310 74
BC102275.1 - 2 - 3 450 74
BT030494.1 - 2 - 3 401 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Gene Ontology Categories:

Function(s) Process(es)

proteasomal ubiquitin-independent protein catabolic process
protein ubiquitination
cellular response to oxidative stress
proteasome-mediated ubiquitin-dependent protein catabolic process
cell redox homeostasis
positive regulation of transcription from RNA polymerase II promoter
cellular response to fluid shear stress
transcription regulatory region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-hrdnk:80/100.66.78.145:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0