This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03253697_m1
See other LIPE GE Assays ›
Gene Symbol:
LIPE
Gene Name:
lipase E, hormone sensitive type
Gene Aliases:
REH, hsl
Chromosome Location:
Chr.18: 50886651 - 50907201 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03253697_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LIPE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080220.1 NP_001073689.1 8 - 9 1691 62
GenBank mRNA EF140760.1 - 8 - 9 1691 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

transcription initiation from RNA polymerase I promoter
termination of RNA polymerase I transcription
cholesterol metabolic process
lipid catabolic process
triglyceride catabolic process
retinol metabolic process
diacylglycerol catabolic process
ether lipid metabolic process
sterol esterase activity
triglyceride lipase activity
hormone-sensitive lipase activity
rRNA primary transcript binding
acylglycerol lipase activity
retinyl-palmitate esterase activity
1,3-diacylglycerol acylhydrolase activity
1,2-diacylglycerol acylhydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0