This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03254530_m1
See other SLC1A4 GE Assays ›
Gene Symbol:
SLC1A4
Gene Name:
solute carrier family 1 member 4
Gene Aliases:
ASCT1
Chromosome Location:
Chr.11: 63379682 - 63409620 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03254530_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC1A4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081577.1 NP_001075046.1 3 - 4 862 57
GenBank mRNA BC133295.1 - 3 - 4 862 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

L-alanine transport
L-cystine transport
L-serine transport
threonine transport
hydroxyproline transport
proline transmembrane transport
cognition
chloride transmembrane transport
chloride channel activity
L-alanine transmembrane transporter activity
L-cystine transmembrane transporter activity
L-proline transmembrane transporter activity
L-serine transmembrane transporter activity
L-threonine transmembrane transporter activity
symporter activity
L-hydroxyproline transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline