Assay Details
| Gene Symbol: SIGIRR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082443.2 | NP_001075912.1 | 3 - 4 | 549 | 64 | |
| GenBank mRNA | BC133493.1 | - | 3 - 4 | 550 | 64 | |
More Information
Set Membership: |
|

SIGIRR
Genomic Map