This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03259195_m1
See other ECE2 GE Assays ›
Gene Symbol:
ECE2
Gene Name:
endothelin converting enzyme 2
Gene Aliases:
EEF1AKMT4, EEF1AKMT4-ECE2
Chromosome Location:
Chr.1: 82894597 - 82927145 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03259195_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ECE2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_174046.4 NP_776471.3 7 - 8 1206 64
NM_177956.3 NP_808871.3 8 - 9 1293 64
NM_177957.3 NP_808872.2 6 - 7 792 64
NM_177958.3 NP_808873.2 7 - 8 879 64
GenBank mRNA AF489573.1 - 7 - 8 1001 64
AF489574.1 - 8 - 9 1088 64
AF489575.1 - 6 - 7 794 64
AF489576.1 - 7 - 8 881 64
U27341.1 - 7 - 8 1001 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

brain development
heart development
cardioblast differentiation
peptide hormone processing
peptidyl-lysine methylation
methylation
metalloendopeptidase activity
methyltransferase activity
protein-lysine N-methyltransferase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4ln64:80/100.66.79.201:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0