This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03259613_g1
See other GPX4 GE Assays ›
Gene Symbol:
GPX4
Gene Name:
glutathione peroxidase 4
Gene Aliases:
PHGPx
Chromosome Location:
Chr.7: 43588660 - 43591107 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03259613_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPX4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001346430.1 NP_001333359.1 3 - 4 511 64
NM_001346431.1 NP_001333360.1 3 - 4 553 64
NM_174770.4 NP_777195.1 3 - 4 440 64
GenBank mRNA AB017534.1 - 3 - 4 334 64
AB098995.1 - 3 - 4 63
BC102957.1 - 3 - 4 316 64
DT835990.1 - 3 - 4 477 64
DT839524.1 - 3 - 4 443 64
DT853625.1 - 3 - 4 415 64
DV828741.1 - 3 - 4 317 64
EH125462.1 - 3 - 4 524 64
EV607600.1 - 3 - 4 511 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

peroxidase

Gene Ontology Categories:

Function(s) Process(es)

chromatin organization
response to oxidative stress
multicellular organism development
spermatogenesis
arachidonic acid metabolic process
lipoxygenase pathway
protein polymerization
oxidation-reduction process
cellular oxidant detoxification
negative regulation of ferroptosis
identical protein binding
phospholipid-hydroperoxide glutathione peroxidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0