This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03259837_m1
See other DGAT2 GE Assays ›
Gene Symbol:
DGAT2
Gene Name:
diacylglycerol O-acyltransferase 2
Gene Aliases:
ARAT
Chromosome Location:
Chr.15: 55160672 - 55193146 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03259837_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DGAT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_205793.2 NP_991362.2 5 - 6 752 57
GenBank mRNA AJ519787.1 - 5 - 6 753 57
AY675174.1 - 2 - 3 281 57
BC105424.1 - 75 57
BT030532.1 - 5 - 6 752 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

glycerol metabolic process
monoacylglycerol biosynthetic process
diacylglycerol biosynthetic process
triglyceride biosynthetic process
lipid storage
low-density lipoprotein particle clearance
long-chain fatty-acyl-CoA metabolic process
cellular triglyceride homeostasis
retinol metabolic process
cholesterol homeostasis
diacylglycerol metabolic process
fatty acid homeostasis
fat pad development
cellular response to oleic acid
regulation of plasma lipoprotein particle levels
2-acylglycerol O-acyltransferase activity
diacylglycerol O-acyltransferase activity
retinol O-fatty-acyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0