This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03259842_g1
See other NAXE GE Assays ›
Gene Symbol:
NAXE
Gene Name:
NAD(P)HX epimerase
Gene Aliases:
AIBP, APOA1BP
Chromosome Location:
Chr.3: 14237518 - 14243767 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03259842_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NAXE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_205796.3 NP_991365.1 3 - 4 439 84
GenBank mRNA AY528250.1 - 3 - 4 440 84
BC114830.1 - 3 - 4 440 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

epimerase/racemase isomerase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

sprouting angiogenesis
regulation of cholesterol efflux
negative regulation of angiogenesis
membrane raft distribution
nicotinamide nucleotide metabolic process
nucleotide binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline