Assay Details
| Gene Symbol: CDK9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014935.2 | NP_001014935.2 | 6 - 7 | 788 | 56 | |
| GenBank mRNA | BC118194.1 | - | 6 - 7 | 788 | 56 | |
| BT020657.1 | - | 6 - 7 | 787 | 56 | ||
| BT021882.1 | - | 6 - 7 | 817 | 56 | ||
More Information
Set Membership: |
|

CDK9
Genomic Map