Assay Details
| Gene Symbol: CHAF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098014.1 | NP_001091483.1 | 11 - 12 | 1603 | 75 | |
| GenBank mRNA | BT030593.1 | - | 11 - 12 | 1603 | 75 | |
More Information
Set Membership: |
|

CHAF1B
Genomic Map