This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03262100_m1
See other SMIM12 GE Assays ›
Gene Symbol:
SMIM12
Gene Name:
small integral membrane protein 12
Gene Aliases:
C3H1orf212
Chromosome Location:
Chr.3: 110835276 - 110839537 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03262100_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SMIM12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001098950.1 NP_001092420.1 1 - 2 73 49
GenBank mRNA BC142001.1 - 1 - 2 112 49
BC142175.1 - 1 - 2 73 49

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC142001.1, BC142175.1, NM_001098950.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0