This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03267916_m1
See other PNLIPRP2 GE Assays ›
Gene Symbol:
PNLIPRP2
Gene Name:
pancreatic lipase related protein 2
Gene Aliases:
PL-RP2
Chromosome Location:
Chr.26: 37019895 - 37039835 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03267916_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PNLIPRP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001105355.2 NP_001098825.1 4 - 5 409 59
GenBank mRNA BC142351.1 - 4 - 5 410 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

cellular defense response
phospholipid catabolic process
response to bacterium
galactolipid catabolic process
intestinal lipid catabolic process
phospholipase activity
triglyceride lipase activity
calcium ion binding
acylglycerol lipase activity
galactolipase activity
1-18:1-2-16:0-monogalactosyldiacylglycerol lipase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0