This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03272350_m1
See other PCED1B GE Assays ›
Gene Symbol:
PCED1B
Gene Name:
PC-esterase domain containing 1B
Gene Aliases:
FAM113B
Chromosome Location:
Chr.5: 33019759 - 33177909 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03272350_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PCED1B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001101207.2 NP_001094677.1 2 - 3 223 62
GenBank mRNA BC147859.1 - 2 - 3 224 62

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC147859.1, NM_001101207.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline