This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03291456_s1
See other PIN4 GE Assays ›
Gene Symbol:
PIN4
Gene Name:
peptidylprolyl cis/trans isomerase, NIMA-interacting 4
Gene Aliases:
Par14
Chromosome Location:
Chr.X: 78475621 - 78484845 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03291456_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PIN4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001105657.1 NP_001099127.1 1 47 73
GenBank mRNA BC149559.1 - 1 47 73

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC149559.1, NM_001105657.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
rRNA processing
DNA binding
peptidyl-prolyl cis-trans isomerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4g6l8:80/100.66.76.118:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0