This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03816110_g1
See other ATOX1 GE Assays ›
Gene Symbol:
ATOX1
Gene Name:
antioxidant 1 copper chaperone
Gene Aliases:
-
Chromosome Location:
Chr.7: 62906883 - 62957606 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03816110_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATOX1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001130758.1 NP_001124230.1 1 - 2 52 79
GenBank mRNA BC102437.1 - 1 - 2 52 79

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC102437.1, NM_001130758.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons

Gene Ontology Categories:

Function(s)

copper ion transport

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-859c6f577-k4jj8:80/100.66.76.192:80.
git-commit: 13ffd1f44fd3b57634547fe5abf56598aa1cdbac
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.44.0-2026.02.07-1.0