This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt04281221_m1
See other EIF1AY GE Assays ›
Gene Symbol:
EIF1AY
Gene Name:
eukaryotic translation initiation factor 1A, Y-linked
Gene Aliases:
-
Chromosome Location:
-
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt04281221_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF1AY
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001145757.1 NP_001139229.1 117 105
GenBank mRNA FJ627277.1 - 108 105

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Bos taurus

Panther Classification:

Molecular Function -

translation initiation factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0