This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt04285075_m1
See other SPTY2D1 GE Assays ›
Gene Symbol:
SPTY2D1
Gene Name:
SPT2 chromatin protein domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.29: 26103453 - 26119994 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt04285075_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SPTY2D1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001205738.1 NP_001192667.1 2 - 3 193 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-flftk:80/100.66.79.227:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0