Assay Details
| Gene Symbol: BORA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205779.2 | NP_001192708.1 | 3 - 4 | 387 | 79 | |
| GenBank mRNA | BC149188.1 | - | 3 - 4 | 343 | 79 | |
More Information
Set Membership: |
|

BORA
Genomic Map