Assay Details
| Gene Symbol: MARCHF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205812.3 | NP_001192741.1 | 16 - 17 | 1460 | 98 | |
| GenBank mRNA | BC119893.1 | - | 6 - 7 | 533 | 98 | |
More Information
Set Membership: |
|

MARCHF6
Genomic Map