Assay Details
| Gene Symbol: TTPAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205931.2 | NP_001192860.1 | 2 - 3 | 552 | 83 | |
| GenBank mRNA | BC149824.1 | - | 1 - 2 | 442 | 83 | |
More Information
Set Membership: |
|

TTPAL
Genomic Map