Assay Details
| Gene Symbol: SDR16C5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205933.2 | NP_001192862.2 | 2 - 3 | 465 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SDR16C5| Gene Symbol: SDR16C5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205933.2 | NP_001192862.2 | 2 - 3 | 465 | 101 | |
Set Membership: |
|