This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt04293923_g1
See other SIGLEC10 GE Assays ›
Gene Symbol:
SIGLEC10
Gene Name:
sialic acid binding Ig like lectin 10
Gene Aliases:
-
Chromosome Location:
Chr.18: 57462616 - 57470264 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt04293923_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SIGLEC10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001206277.1 NP_001193206.1 4 - 5 933 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin superfamily cell adhesion molecule

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline