This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt04299713_m1
See other EEF1D GE Assays ›
Gene Symbol:
EEF1D
Gene Name:
eukaryotic translation elongation factor 1 delta
Gene Aliases:
-
Chromosome Location:
Chr.14: 1113581 - 1126356 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt04299713_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EEF1D
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001206620.2 NP_001193549.1 3 - 4 485 91
NM_001289966.1 NP_001276895.1 5 - 6 1567 91
GenBank mRNA BC105375.1 - 4 - 5 1290 91
BT030508.1 - 3 - 4 245 91
EV683636.1 - 3 - 4 457 91
KC190038.1 - 3 - 4 1478 91
KC190039.1 - 3 - 4 485 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

translation elongation factor

Gene Ontology Categories:

Process(es)

translation elongation factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0