This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt04301238_m1
See other UCP1 GE Assays ›
Gene Symbol:
UCP1
Gene Name:
uncoupling protein 1
Gene Aliases:
-
Chromosome Location:
Chr.17: 17164837 - 17171147 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt04301238_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UCP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001166528.1 NP_001160000.1 2 - 3 335 74
GenBank mRNA X14064.1 - 276 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

diet induced thermogenesis
regulation of transcription from RNA polymerase II promoter
response to temperature stimulus
response to nutrient levels
cellular response to hormone stimulus
cellular response to reactive oxygen species
brown fat cell differentiation
cellular response to cold
cellular response to fatty acid
positive regulation of cold-induced thermogenesis
hydrogen ion transmembrane transport
regulation of reactive oxygen species biosynthetic process
mitochondrial transmembrane transport
adaptive thermogenesis
oxidative phosphorylation uncoupler activity
purine ribonucleotide binding
long-chain fatty acid binding
cardiolipin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nbrrd:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0