This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt04301492_m1
See other OGFOD1 GE Assays ›
Gene Symbol:
OGFOD1
Gene Name:
2-oxoglutarate and iron dependent oxygenase domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.18: 24153095 - 24184693 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt04301492_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OGFOD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001166613.1 NP_001160085.1 3 - 4 414 65
GenBank mRNA BC104499.1 - 3 - 4 337 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

regulation of translational termination
cell proliferation
protein hydroxylation
stress granule assembly
oxidation-reduction process
iron ion binding
L-ascorbic acid binding
peptidyl-proline dioxygenase activity
peptidyl-proline 3-dioxygenase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0