Assay Details
| Gene Symbol: NADK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256553.2 | NP_001243482.1 | 1 - 2 | 408 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NADK2| Gene Symbol: NADK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256553.2 | NP_001243482.1 | 1 - 2 | 408 | 58 | |
Set Membership: |
|